Anti-MEOX 2 Mouse Monoclonal Antibody [clone: 5E1.]
Catalog # 103330-810
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:MEOX 2
- Clonality:Monoclonal
- Clone:5E1.
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:4223
- Antigen Synonyms:mesenchyme homeo box 2 (growth arrest-specific homeo box)|Growth arrest-specific homeobox|mesenchyme homeobox 2GAX|homeobox protein MOX-2|growth arrest-specific homeo box|MOX2mesenchyme homeobox 2 (growth arrest-specific homeo box)
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:MEOX2 (AAH17021, 1 a.a. - 303 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MEHPLFGCLRSPHATAQGLHPFSQSSLALHGRSDHMSYPELSTSSSSCIIAGYPNEEGMFASQHHRGHHHHHHHHHHHHQQQQHQALQTNWHLPQMSSPPSAARHSLCLQPDSGGPPELGSSPPVLCSNSSSLGSSTPTGAACAPGDYGRQALSPAEAEKRSGGKRKSDSSDSQEGNYKSEVNSKPRKERTAFTKEQIRELEAEFAHHNYLTRLRRYEIAVNLDLTERQVKVWFQNRRMKWKRVKGGQQGAAAREKELVNVKKGTLLPSELSGIGAATLQQTGDSIANEDSHDSDHSSEHAHL
- Purification:IgG purified
- Cat. No.:103330-810
- Supplier no.:H00004223-M08
Specifications
About this item
The MEOX 2 Antibody (5E1.) from Novus Biologicals is a mouse monoclonal antibody to MEOX 2. This antibody reacts with human, mouse, rat. The MEOX 2 Antibody (5E1.) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: MEOX 2
Clonality: Monoclonal
Clone: 5E1.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat