Anti-MDFI Mouse Monoclonal Antibody [clone: 3B4]
Catalog # 103330-642
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:MDFI
- Clonality:Monoclonal
- Clone:3B4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:4188
- Antigen Synonyms:Myogenic repressor I-mf|I-mfa|I-MF|MyoD family inhibitor|inhibitor of MyoD family a
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:MDFI (NP_005577, 33 a.a. - 110 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PGLEVVTGSTHPAEAAPEEGSLEEAATPMPQGNGPGIPQGLDSTDLDVPTEAVTCQPQGNPLGCTPLLPNDSGHPSEL
- Purification:IgG purified
- Cat. No.:103330-642
- Supplier no.:H00004188-M07
Specifications
About this item
The MDFI Antibody (3B4) from Novus Biologicals is a mouse monoclonal antibody to MDFI. This antibody reacts with human. The MDFI Antibody (3B4) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: MDFI
Clonality: Monoclonal
Clone: 3B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human