To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:MCM7
- Clonality:Monoclonal
- Clone:2068C
- Conjugation:DyLight® 650
- Flow Cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:4176
- Antigen Synonyms:CDC47P85MCM|EC 3.6.4.12|minichromosome maintenance deficient 7|minichromosome maintenance deficient (S. cerevisiae) 7|P1.1-MCM3|P1CDC47|CDC47 homolog|DNA replication licensing factor MCM7|minichromosome maintenance complex component 7|homolog of S. cerevisiae Cdc47|MCM2PNAS146|MCM7 minichromosome maintenance deficient 7 (S. cerevisiae)
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Partial recombinant human MCM-7 protein (amino acids 188-328) [UniProt P33993]
- Purification:Protein A or G purified
- Cat. No.:103295-126
- Supplier no.:MAB92171C
The MCM7 Antibody (2068C) [DyLight 650] from Novus Biologicals is a rabbit monoclonal antibody to MCM7. This antibody reacts with human. The MCM7 Antibody (2068C) [DyLight 650] has been validated for the following applications: Western Blot, Flow Cytometry.
Type: Primary
Antigen: MCM7
Clonality: Monoclonal
Clone: 2068C
Conjugation: DyLight 650
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human