Anti-MCF2L Mouse Monoclonal Antibody [clone: 1D11]
103341-146
- Antibody Type:Primary
- Antigen Symbol:MCF2L
- Clonality:Monoclonal
- Clone:1D11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:23263
- Antigen Synonyms:DBS|FLJ12122|KIAA0362ARHGEF14|MCF2 transforming sequence-like protein|MCF.2 cell line derived transforming sequence-like|DBL's big sister|OST|MCF2-transforming sequence-like protein|guanine nucleotide exchange factor DBS
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:MCF2L (AAH20208, 220 a.a. ~ 320 a.a) partial recombinant protein with GST tag.GTELAETELPNDVQSTSSVLCAHTEKKDKAKEDLRLALKEGHSVLESLRELQAEGSEPSVNQDQLDNQATVQRLLAQLNETEAAFDEFWAKHQQKLEQCL
- Purification:IgG purified
- Cat. No.:103341-146
- Supplier no.:H00023263-M01
The MCF2L Antibody (1D11) from Novus Biologicals is a mouse monoclonal antibody to MCF2L. This antibody reacts with human. The MCF2L Antibody (1D11) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: MCF2L
Clonality: Monoclonal
Clone: 1D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human