Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:MCAM/CD146
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:4162
- Antigen Synonyms:Melanoma-associated antigen MUC18|melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen|Cell surface glycoprotein P1H12|cell surface glycoprotein MUC18|Melanoma-associated antigen A32|MUC18Gicerin|CD146 antigen|melanoma adhesion molecule|CD146
- Storage Buffer:PBS (pH 7.0), 1.0% BSA and 20% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the C-terminus region of human CD146. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Cat. No.:102706-650
- Supplier no.:NBP2-15778
Specifications
About this item
The MCAM / CD146 Antibody from Novus Biologicals is a rabbit polyclonal antibody to MCAM / CD146. This antibody reacts with human, mouse, rat. The MCAM / CD146 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: MCAM/CD146
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat