Anti-MAT2A Rabbit Polyclonal Antibody
103287-638
- Antibody Type:Primary
- Antigen Symbol:MAT2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:4144
- Antigen Synonyms:Methionine adenosyltransferase II|alpha|MAT 2|MATA2SAMS2adoMet synthetase 2|Methionine adenosyltransferase 2|methionine adenosyltransferase II|MAT-II|AMS2|S-adenosylmethionine synthase isoform type-2|S-adenosylmethionine synthetase isoform type-2|MATII|EC 2.5.1.6|AdoMet synthase 2
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1 mg/ml
- Immunogen:The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Methionine Adenosyltransferase II, Alpha using the numbering given in entry NP_005902.1 (GeneID 4144).
- Purification:Immunogen affinity purified
- Cat. No.:103287-638
- Supplier no.:NBP2-32283
The MAT2A Antibody from Novus Biologicals is a rabbit polyclonal antibody to MAT2A. This antibody reacts with human, mouse. The MAT2A Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: MAT2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse