Anti-MAP7 Mouse Polyclonal Antibody
Catalog # 103336-122
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:MAP7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:9053
- Antigen Synonyms:MAP-7|Epithelial microtubule-associated protein of 115 kDa|E-MAP-115dJ325F22.2 (microtubule-associated protein 7 (EMAP115|E-MAP-115))|ensconsin|microtubule-associated protein 7EMAP115
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:MAP7 (NP_003971.1, 1 a.a. - 749 a.a.) full-length human protein. MAELGAGGDGHRGGDGAVRSETAPDSYKVQDKKNASSRPASAISGQNNNHSGNKPDPPPVLRVDDRQRLARERREEREKQLAAREIVWLEREERARQHYEKHLEERKKRLEEQRQKEERRRAAVEEKRRQRLEEDKERHEAVVRRTMERSQKPKQKHNRWSWGGSLHGSPSIHSADPDRRSVSTMNLSKYVDPVISKRLSSSSATLLNSPDRARRLQLSPWESSVVNRLLTPTHSFLARSKSTAALSGEAASCSPIIMPYKAAHSRNSMDRPKLFVTPPEGSSRRRIIHGTASYKKERERENVLFLTSGTRRAVSPSNPKARQPARSRLWLPSKSLPHLPGTPRPTSSLPPGSVKAAPAQVRPPSPGNIRPVKREVKVEPEKKDPEKEPQKVANEPSLKGRAPLVKVEEATVEERTPAEPEVGPAAPAMAPAPASAPAPASAPAPAPVPTPAMVSAPSSTVNASASVKTSAGTTDPEEATRLLAEKRRLAREQREKEERERREQEELERQKREELAQRVAEERTTRREEESRRLEAEQAREKEEQLQRQAEERALREREEAERAQRQKEEEARVREEAERVRQEREKHFQREEQERLERKKRLEEIMKRTRRTEATDKKTSDQRNGDIAKGALTGGTEVSALPCTTNAPGNGKPVGSPHVVTSHQSKVTVESTPDLEKQPNENGVSVQNENFEEIINLPIGSKPSRLDVTNSESPEIPLNPILAFDDEGTLGPLPQVDGVQTQQTAEVI
- Purification:Protein G purified
- Cat. No.:103336-122
- Supplier no.:H00009053-B01P
Specifications
About this item
The MAP7 Antibody from Novus Biologicals is a mouse polyclonal antibody to MAP7. This antibody reacts with human. The MAP7 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: MAP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human