Anti-MAF1 Mouse Polyclonal Antibody
Catalog # 103345-116
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:MAF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 ug
- Gene ID:84232
- Antigen Synonyms:MGC39758|repressor of RNA polymerase III transcription MAF1 homolog|MGC31779|homolog of yeast MAF1|DKFZp586G1123|MGC20332|MAF1 homolog (S. cerevisiae)
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:MAF1 (AAH31273.1, 1 a.a. ~ 256 a.a) full-length human protein.
- Purification:Protein G purified
- Cat. No.:103345-116
- Supplier no.:H00084232-B01P
Specifications
About this item
The MAF1 Antibody from Novus Biologicals is a mouse polyclonal antibody to MAF1. This antibody reacts with human. The MAF1 Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: MAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human