Anti-LZTS2 Rabbit Polyclonal Antibody
103258-922
- Antibody Type:Primary
- Antigen Symbol:LZTS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:84445
- Antigen Synonyms:KIAA1813leucine zipper putative tumor suppressor 2|leucine zipper|LAPSER1Protein LAPSER1|putative tumor suppressor 2|hLZTS2
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:EARATLRVSEGRARGLQEAARARELELEACSQELQRHRQEAEQLREKAGQLDAEAAGLREPPVPPATADPFLLAESDEAKVQRAA
- Purification:Immunogen affinity purified
- Cat. No.:103258-922
- Supplier no.:NBP1-82892
The LZTS2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to LZTS2. This antibody reacts with human. The LZTS2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: LZTS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human