Anti-LYAG/GAA Mouse Monoclonal Antibody [clone: 3C6]
Catalog # 103326-886
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:LYAG/GAA
- Clonality:Monoclonal
- Clone:3C6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:2548
- Antigen Synonyms:lysosomal alpha-glucosidase|alpha|Aglucosidase alfa|LYAG|glucosidase|EC 3.2.1.20|Acid maltase|acid
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:GAA (AAH40431 851 a.a. - 952 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GEARGELFWDDGESLEVLERGAYTQVIFLARNNTIVNELVRVTSEGAGLQLQKVTVLGVATAPQQVLSNGVPVSNFTYSPDTKVLDICVSLLMGEQFLVSWC
- Purification:IgG purified
- Cat. No.:103326-886
- Supplier no.:H00002548-M01
Specifications
About this item
The LYAG / GAA Antibody (3C6) from Novus Biologicals is a mouse monoclonal antibody to LYAG / GAA. This antibody reacts with human. The LYAG / GAA Antibody (3C6) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: LYAG/GAA
Clonality: Monoclonal
Clone: 3C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human