Anti-LRRC49 Rabbit Polyclonal Antibody
Catalog # 103286-472
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:LRRC49
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Gene ID:54839
- Antigen Synonyms:leucine rich repeat containing 49|FLJ20156|Tubulin polyglutamylase complex subunit 4|PGs4|leucine-rich repeat-containing protein 49
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids: SHVDNLNGLDSLTELNLRHNQITFVRDVDNLPCLQHLFLSFNNISSFDSV SCLADSSSLSDITFDGNPIAQESWYKHTVLQNMMQLRQLDMKRITEEERR MASVLAKKEEEKKRESHKQSLLKEKKRLTINNVARQWDLQQQRVA
- Purification:Immunogen affinity purified
- Cat. No.:103286-472
- Supplier no.:NBP2-14202
Specifications
About this item
The LRRC49 Antibody from Novus Biologicals is a rabbit polyclonal antibody to LRRC49. This antibody reacts with human. The LRRC49 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: LRRC49
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human