Anti-LRRC17 Mouse Polyclonal Antibody
Catalog # 103338-480
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:LRRC17
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:10234
- Antigen Synonyms:37 kDa leucine-rich repeat (LRR) protein|P37NBH_RG318M05.3|leucine-rich repeat-containing protein 17|p37NB|leucine rich repeat containing 17
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:LRRC17 (NP_001026862.1, 1 a.a. ~ 441 a.a) full-length human protein.
- Purification:Protein G purified
- Cat. No.:103338-480
- Supplier no.:H00010234-B01P
Specifications
About this item
The LRRC17 Antibody from Novus Biologicals is a mouse polyclonal antibody to LRRC17. This antibody reacts with human. The LRRC17 Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: LRRC17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human