Anti-LRAT Mouse Monoclonal Antibody [clone: 4E12]
Catalog # 103336-462
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:LRAT
- Clonality:Monoclonal
- Clone:4E12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:9227
- Antigen Synonyms:Phosphatidylcholine--retinol O-acyltransferase|lecithin retinol acyltransferase|LCA14|MGC33103|lecithin retinol acyltransferase (phosphatidylcholine--retinolO-acyltransferase)|EC 2.3.1.135
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:LRAT (NP_004735, 33 a.a. ~ 133 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Purification:IgG purified
- Cat. No.:103336-462
- Supplier no.:H00009227-M05
Specifications
About this item
The LRAT Antibody (4E12) from Novus Biologicals is a mouse monoclonal antibody to LRAT. This antibody reacts with human. The LRAT Antibody (4E12) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: LRAT
Clonality: Monoclonal
Clone: 4E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human