Anti-LIMK-2 Mouse Monoclonal Antibody [clone: 2H2-E11]
Catalog # 103330-156
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:LIM kinase 2
- Antigen Symbol:LIMK-2
- Clonality:Monoclonal
- Clone:2H2-E11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:3985
- Antigen Synonyms:LIM domain kinase 2|EC 2.7.11.1|LIMK-2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:LIMK2 (AAH13051.1, 1 a.a. - 686 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MGSYLSVPAYFTSRDLFRCSECQDSLTNWYYEKDGKLYCPKDYWGKFGEFCHGCSLLMTGPFMVAGEFKYHPECFACMSCKVIIEDGDAYALVQHATLYCGKCHNEVVLAPMFERLSTESVQEQLPYSVTLISMPATTEGRRGFSVSVESACSNYATTVQVKEVNRMHISPNNRNAIHPGDRILEINGTPVRTLRVEEVEDAISQTSQTLQLLIEHDPVSQRLDQLRLEARLAPHMQNAGHPHALSTLDTKENLEGTLRRRSLRRSNSISKSPGPSSPKEPLLFSRDISRSESLRCSSSYSQQIFRPCDLIHGEVLGKGFFGQAIKVTHKATGKVMVMKELIRCDEETQKTFLTEVKVMRSLDHPNVLKFIGVLYKDKKLNLLTEYIEGGTLKDFLRSMDPFPWQQKVRFAKGIASGMAYLHSMCIIHRDLNSHNCLIKLDKTVVVADFGLSRLIVEERKRAPMEKATTKKRTLRKNDRKKRYTVVGNPYWMAPEMLNGKSYDETVDIFSFGIVLCEIIGQVYADPDCLPRTLDFGLNVKLFWEKFVPTDCPPAFFPLAAICCRLEPESRAPPGAAGEGPGCADDEGPVRRQGKVTIKYDPKELRKHLNLEEWILEQLTRLYDCQEEEISELEIDVDELLDMESDDAWASRVKELLVDCYKPTEAFISGLLDKIRAMQKLSTPQKK
- Purification:IgG purified
- Cat. No.:103330-156
- Supplier no.:H00003985-M01
Specifications
About this item
The LIM kinase 2 Antibody (2H2-E11) from Novus Biologicals is a mouse monoclonal antibody to LIM kinase 2. This antibody reacts with human. The LIM kinase 2 Antibody (2H2-E11) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: LIMK-2
Clonality: Monoclonal
Clone: 2H2-E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human