Anti-Lefty-B Mouse Monoclonal Antibody [clone: 2E10]
Catalog # 103339-322
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:Lefty-B
- Clonality:Monoclonal
- Clone:2E10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:10637
- Antigen Synonyms:Left-right determination factor B|LEFTBProtein lefty-B|left-right determination factor 1|LEFTYBProtein lefty-1|left-right determination|factor B
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:LEFTY1 (AAH27883, 1 a.a. ~ 367 a.a) full length recombinant protein with GST tag.MQPLWLCWALWVLPLASPGAALTGEQLLGSLLRQLQLKEVPT LDRADMEELVIPTHVRAQYVALLQRSHGDRSRGKRFSQSFRE VAGRFLALEASTHLLVFGMEQRLPPNSELVQAVLRLFQEPVP KAALHRHGRLSPRSARARVTVEWLRVRDDGSNRTSLIDSRLV SVHESGWKAFDVTEAVNFWQQLSRPRQPLLLQVSVQREHLGP LASGAHKLVRFASQGAPAGLGEPQLELHTLDLGDY
- Purification:IgG purified
- Cat. No.:103339-322
- Supplier no.:H00010637-M03
Specifications
About this item
The Lefty-B Antibody (2E10) from Novus Biologicals is a mouse monoclonal antibody to Lefty-B. This antibody reacts with human. The Lefty-B Antibody (2E10) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: Lefty-B
Clonality: Monoclonal
Clone: 2E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human