Anti-Kv3.3 Mouse Monoclonal Antibody [clone: 1C1]
Catalog # 103329-786
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:Kv3.3
- Clonality:Monoclonal
- Clone:1C1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:3748
- Antigen Synonyms:SCA13|potassium voltage-gated channel|KV3.3|Voltage-gated potassium channel subunit Kv3.3|potassium voltage-gated channel subfamily C member 3|Shaw-related voltage-gated potassium channel protein 3|spinocerebellar ataxia 13|Shaw-related subfamily|member 3|KSHIIID|voltage-gated potassium channel protein KV3.3
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:KCNC3 (NP_004968 671 a.a. - 757 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ALAHEDCPAIDQPAMSPEDKSPITPGSRGRYSRDRACFLLTDYAPSPDGSIRKATGAPPLPPQDWRKPGPPSFLPDLNANAAAWISP
- Purification:IgG purified
- Cat. No.:103329-786
- Supplier no.:H00003748-M01
Specifications
About this item
The Kv3.3 Antibody (1C1) from Novus Biologicals is a mouse monoclonal antibody to Kv3.3. This antibody reacts with human, mouse. The Kv3.3 Antibody (1C1) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: Kv3.3
Clonality: Monoclonal
Clone: 1C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse