Anti-KLF6 Mouse Monoclonal Antibody [clone: 3C4]
Catalog # 103324-200
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:KLF6
- Clonality:Monoclonal
- Clone:3C4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 μg
- Gene ID:1316
- Antigen Synonyms:suppression of tumorigenicity 12 (prostate)|CPBPZF9|Transcription factor Zf9|protooncogene B-cell derived 1|PAC1|Kruppel-like factor 6|COPEBCBA1|GC-rich binding factor|Kruppel-like zinc finger protein Zf9|core promoter element binding protein|GC-rich sites-binding factor GBF|EC 6.1.1.15|B-cell-derived protein 1|Krueppel-like factor 6|EC 2.1.1.43|ST12|Proto-oncogene BCD1|Suppressor of tumorigenicity 12 protein|BCD1Zf9|GBF|DKFZp686N0199|Core promoter element-binding protein
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:KLF6 (NP_001291, 1 a.a. - 100 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MDVLPMCSIFQELQIVHETGYFSALPSLEEYWQQTCLELERYLQSEPCYVSASEIKFDSQEDLWTKIILAREKKEESELKISSSPPEDTLISPSFCYNLE
- Purification:IgG purified
- Cat. No.:103324-200
- Supplier no.:H00001316-M02
Specifications
About this item
The KLF6 Antibody (3C4) from Novus Biologicals is a mouse monoclonal antibody to KLF6. This antibody reacts with human. The KLF6 Antibody (3C4) has been validated for the following applications: ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: KLF6
Clonality: Monoclonal
Clone: 3C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human