Anti-KCNMB4 Mouse Monoclonal Antibody [clone: 1G7]
Catalog # 103342-830
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:KCNMB4
- Clonality:Monoclonal
- Clone:1G7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 μg
- Gene ID:27345
- Antigen Synonyms:K(VCA)beta-4|potassium large conductance calcium-activated channel|calcium-activated potassium channel subunit beta-4|Calcium-activated potassium channel|large conductance calcium-dependent potassium ion channel beta 4 subunit|slo-beta-4|subfamily M|subfamily M subunit beta-4|Charybdotoxin receptor subunit beta-4|BK channel subunit beta-4|beta member4|Maxi K channel subunit beta-4|hbeta4|BKbeta4
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:KCNMB4 (AAH50621, 1 a.a. ~ 211 a.a) full length recombinant protein with GST tag.MAKLRVAYEYTEAEDKSIRLGLFLIISGVVSLFIFGFCWLSPALQDLQATEANCTVLSVQQIGEVFECTFTCGADCRGTSQYPCVQVYVNNSESNSRALLHSDEHQLLTNPKCSYIPPCKRENQKNLESVMNWQQYWKDEIGSQPFTCYFNQHQRPDDVLLHRTHDEIVLLHCFLWPLVTFVVGVLIVVLTICAKSLAVKAEAMKKRKFS
- Purification:IgG purified
- Cat. No.:103342-830
- Supplier no.:H00027345-M01
Specifications
About this item
The KCNMB4 Antibody (1G7) from Novus Biologicals is a mouse monoclonal antibody to KCNMB4. This antibody reacts with human. The KCNMB4 Antibody (1G7) has been validated for the following applications: ELISA.
Type: Primary
Antigen: KCNMB4
Clonality: Monoclonal
Clone: 1G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human