Anti-JMY Mouse Monoclonal Antibody [clone: 2D7]
Catalog # 103346-860
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:JMY
- Clonality:Monoclonal
- Clone:2D7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:133746
- Antigen Synonyms:junction-mediating and regulatory protein|FLJ37870|WHDC1L3|p53 cofactor|junction-mediating and -regulatory protein|MGC163496|junction mediating and regulatory protein|WAS protein homology region 2 domain containing 1-like 3
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:JMY (NP_689618, 535 a.a. ~ 635 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.SPMDEVLASLKRGSFHLKKVEQRTLPPFPDEDDSNNILAQIRKGVKLKKVQKDVLRESFTLLPDTDPLTRSIHEALRRIKEASPESEDEEEALPCTDWEN
- Purification:IgG purified
- Cat. No.:103346-860
- Supplier no.:H00133746-M01
Specifications
About this item
The JMY Antibody (2D7) from Novus Biologicals is a mouse monoclonal antibody to JMY. This antibody reacts with human. The JMY Antibody (2D7) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: JMY
Clonality: Monoclonal
Clone: 2D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human