Anti-ITGA8 Mouse Monoclonal Antibody [clone: 2G7]
Catalog # 103335-068
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Integrin alpha 8
- Antigen Symbol:ITGA8
- Clonality:Monoclonal
- Clone:2G7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:8516
- Antigen Synonyms:integrin alpha-8|alpha 8|integrin
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:ITGA8 (NP_003629 836 a.a. - 935 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SDTILEVGWPFSARDEFLLYIFHIQTLGPLQCQPNPNINPQDIKPAASPEDTPELSAFLRNSTIPHLVRKRDVHVVEFHRQSPAKILNCTNIECLQISCA
- Purification:IgG purified
- Cat. No.:103335-068
- Supplier no.:H00008516-M02
Specifications
About this item
The Integrin alpha 8 Antibody (2G7) from Novus Biologicals is a mouse monoclonal antibody to Integrin alpha 8. This antibody reacts with human. The Integrin alpha 8 Antibody (2G7) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: ITGA8
Clonality: Monoclonal
Clone: 2G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human