Anti-INHAT-1 Rabbit Polyclonal Antibody
Catalog # 103358-898
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:INHAT-1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.025 mg
- Antigen Synonyms:TAF-1b|TAF-1a
- Storage Buffer:0.2 ml PBS and 0.1 mg/ml BSA
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:This antibody was raised against synthetic peptides of amino acids 66-81 and 135-151 of human INHAT-1. Sequence homology is 100% in mouse and rat.
- Purification:Protein G purified
- Cat. No.:103358-898
Specifications
About this item
The INHAT-1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to INHAT-1. This antibody reacts with human, mouse, rat. The INHAT-1 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: INHAT-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat