Anti-IL-1 RI Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: JAMA-147]
103407-032
- Antibody Type:Primary
- Antigen Symbol:IL-1 RI
- Clonality:Monoclonal
- Clone:JAMA-147
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Armenian hamster
- Isotype:IgG
- Reactivity:Mouse
- Size:100 μl
- Gene ID:3554
- Antigen Synonyms:IL1RT1|Interleukin 1 Receptor 1|p80|IL-1RT1|IL1R|IL1R1|interleukin-1 receptor type 1|CD121 antigen-like family member A|IL-1RT-1|CD121a antigen|interleukin receptor 1|type I|interleukin 1 receptor
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Extracellular domain of murine IL-1R.
- Purification:Protein G purified
- Cat. No.:103407-032
The IL-1 RI Antibody (JAMA-147) [PE] from Novus Biologicals is a armenian hamster monoclonal antibody to IL-1 RI. This antibody reacts with mouse. The IL-1 RI Antibody (JAMA-147) [PE] has been validated for the following applications: Flow Cytometry.
Type: Primary
Antigen: IL-1 RI
Clonality: Monoclonal
Clone: JAMA-147
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse