Anti-IKK beta Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 10A9B6]
Catalog # 103359-520
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:IKK beta
- Clonality:Monoclonal
- Clone:10A9B6
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:3551
- Antigen Synonyms:MGC131801|I-kappa-B kinase 2|IKKBFLJ40509|IKK-B|NFKBIKBFLJ33771|IkBKB|inhibitor of kappa light polypeptide gene enhancer in B-cells|kinase beta|I-kappa-B-kinase beta|Nuclear factor NF-kappa-B inhibitor kinase beta|EC 2.7.11|IKK-betaFLJ36218|IKK2FLJ38368|EC 2.7.11.10|inhibitor of nuclear factor kappa-B kinase subunit beta
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Full-length human IKKb/IKK2 recombinant protein was used as immunogen (NP_001547).
- Purification:Protein G purified
- Cat. No.:103359-520
Specifications
About this item
The IKK beta Antibody (10A9B6) [HRP] from Novus Biologicals is a mouse monoclonal antibody to IKK beta. This antibody reacts with human, mouse. The IKK beta Antibody (10A9B6) [HRP] has been validated for the following applications: Western Blot.
Type: Primary
Antigen: IKK beta
Clonality: Monoclonal
Clone: 10A9B6
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse