Anti-IFGR2 Hamster Monoclonal Antibody (DyLight 680) [clone: RM0252-7A25]
Catalog # 103409-592
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Interferon gamma R2
- Antigen Symbol:IFGR2
- Clonality:Monoclonal
- Clone:RM0252-7A25
- Conjugation:DyLight® 680
- Flow Cytometry:Yes
- Host:Hamster
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:3460
- Antigen Synonyms:interferon gamma receptor accessory factor-1|IFN-gamma receptor 2|Interferon gamma transducer 1|interferon gamma receptor beta chain|interferon gamma receptor 2 (interferon gamma transducer 1)|IFGR2|AF-1interferon-gamma receptor beta chain precursor10IFNGT1Interferon gamma receptor accessory factor 1|IFN-gamma-R2|interferon gamma receptor 2
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Mouse recombinant protein of IFNGR2, extracellular domain
- Purification:>95% pure by SDS-PAGE
- Cat. No.:103409-592
Specifications
About this item
The IFN-gamma R2 Antibody (RM0252-7A25) [DyLight 680] from Novus Biologicals is a hamster monoclonal antibody to IFN-gamma R2. This antibody reacts with mouse. The IFN-gamma R2 Antibody (RM0252-7A25) [DyLight 680] has been validated for the following applications: Western Blot, Flow Cytometry.
Type: Primary
Antigen: IFGR2
Clonality: Monoclonal
Clone: RM0252-7A25
Conjugation: DyLight 680
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse