To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:HSPA8/HSC71/Hsc70
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:3312
- Antigen Synonyms:heat shock cognate 71 kDa protein|heat shock cognate protein|HSC71|NIP71|HSPA10|71-kDa|N-myristoyltransferase inhibitor protein 71|LPS-associated protein 1|MGC131511|HSC54|MGC29929|Heat shock 70 kDa protein 8|heat shock 70kD protein 8|HSP71|HSC70heat shock 70kd protein 10|lipopolysaccharide-associated protein 1|LAP1|heat shock cognate protein 54|heat shock 70kDa protein 8|HSP73constitutive heat shock protein 70
- Storage Buffer:0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:0.25 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human HSC70. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Cat. No.:102708-722
- Supplier no.:NBP2-16882
Specifications
About this item
The HSPA8 / HSC71 / Hsc70 Antibody from Novus Biologicals is a rabbit polyclonal antibody to HSPA8 / HSC71 / Hsc70. This antibody reacts with human, mouse. The HSPA8 / HSC71 / Hsc70 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: HSPA8/HSC71/Hsc70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse