Anti-HLA DRB1 Mouse Monoclonal Antibody (Allophycocyanin) [clone: DA2]
103357-390
- Antibody Type:Primary
- Antigen Symbol:HLA DRB1
- Clonality:Monoclonal
- Clone:DA2
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 μl
- Epitope:1st domain, monomorphic.
- Gene ID:3123
- Antigen Synonyms:Clone P2-beta-3|DR-8|DW2.2/DR2.2|MHC class II antigen DRB1*12|DR16|FLJ76359|DR beta 1|DR4|DR-13|MHC class II antigen DRB1*11|HLA-DR1B|lymphocyte antigen DRB1|MHC class II antigen HLA-DR13|DRB1|DR-1 beta chain|MHC class II antigen DRB1*9|major histocompatibility complex|DRw8|MHC class II antigen DRB1*13|MHC class II antigen DRB1*10|DR13|class II|MHC class II HLA-DR beta 1 chain|DR7|DR-9|DR-12|HLA-DRB|DR8|FLJ75017|DR14|DR-4|DR-14|DR-16|MHC class II antigen DRB1*15|DR1|HLA-DR-beta 1|MHC class II antigen DRB1*7|DRw11|DR9|MHC class II antigen DRB1*3|DRw10|HLA-DRB1*|HLA-DRB2|HLA class II histocompatibility antigen|DR-7|MHC class II antigen DRB1*1|DR-1|MHC class II HLA-DR-beta cell surface gly|MHC class II antigen DRB1*14|leucocyte antigen DR beta 1 chain|MHC class II antigen DRB1*8|leucocyte antigen DRB1|HLA class II antigen beta chain|DR-5|DR5|MHC class II antigen DRB1*16|MHC class II antigen DRB1*4|human leucocyte antigen DRB1|DR12
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:The membranes of LKT cells (a B lymphoblastoid cell line)
- Purification:Protein A or G purified
- Cat. No.:103357-390
The HLA DRB1 Antibody (DA2) [Allophycocyanin] from Novus Biologicals is a mouse monoclonal antibody to HLA DRB1. This antibody reacts with human. The HLA DRB1 Antibody (DA2) [Allophycocyanin] has been validated for the following applications: Flow Cytometry.
Type: Primary
Antigen: HLA DRB1
Clonality: Monoclonal
Clone: DA2
Conjugation: Allophycocyanin
Epitope: 1st domain, monomorphic.
Host: Mouse
Isotype: IgG1
Reactivity: Human