Anti-HIV-1 gp41 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 417D3.08/ clone 2]
Catalog # 103319-064
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:HIV-1 gp41
- Clonality:Monoclonal
- Clone:417D3.08/ clone 2
- Conjugation:Alexa Fluor® 488
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Virus,Human
- Size:100 μg
- Antigen Synonyms:gp41|Glycoprotein 41|Transmembrane protein|Human Immunodeficiency Virus Type 1 gp41|Human immunodeficiency virus 1|Env|TM
- Storage Buffer:Antibody in 100 ug in 200 ul/ 50 ug in 100 ul PBS and 50% Glycerol
- Storage Temperature:Store at -20C in the dark. Avoid freeze-thaw cycles.
- Immunogen:HR1-PID-HR2-transfected HEK 293 cells (HIV-1, 92BR025, Clade C)
- Purification:Ion exchange chromatography
- Cat. No.:103319-064
- Supplier no.:DDX1305A488
Specifications
About this item
The HIV-1 gp41 Antibody (417D3.08 / clone 2) [Alexa Fluor« 488] from Novus Biologicals is a mouse monoclonal antibody to HIV-1 gp41. This antibody reacts with human, virus. The HIV-1 gp41 Antibody (417D3.08 / clone 2) [Alexa Fluor« 488] has been validated for the following applications: Flow Cytometry, ELISA.
Type: Primary
Antigen: HIV-1 gp41
Clonality: Monoclonal
Clone: 417D3.08/ clone 2
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Virus