Anti-HIPK2 Mouse Monoclonal Antibody [clone: 1D8]
Catalog # 103342-950
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:HIPK2
- Clonality:Monoclonal
- Clone:1D8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:28996
- Antigen Synonyms:PRO0593|DKFZp686K02111|hHIPk2|homeodomain interacting protein kinase 2|EC 2.7.11.1|FLJ23711|homeodomain-interacting protein kinase 2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:HIPK2 (NP_073577, 213 a.a. ~ 293 a.a) full length recombinant protein with GST tag.
- Purification:IgG purified
- Cat. No.:103342-950
- Supplier no.:H00028996-M08
Specifications
About this item
The HIPK2 Antibody (1D8) from Novus Biologicals is a mouse monoclonal antibody to HIPK2. This antibody reacts with human. The HIPK2 Antibody (1D8) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: HIPK2
Clonality: Monoclonal
Clone: 1D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human