To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:HIF-2 alpha/EPAS1
- Clonality:Monoclonal
- Clone:ep190b
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Hamster,Human,Rat,Mouse
- Size:100 μl
- Gene ID:2034
- Antigen Synonyms:Basic-helix-loop-helix-PAS protein MOP2|HIF2A|HIF2AHIF-2-alpha|Class E basic helix-loop-helix protein 73|hif2a angiogenesis|HIF-1alpha-like factor|bHLHe73ECYT4|endothelial PAS domain-containing protein 1|HLFHIF-1-alpha-like factor|EPAS-1|HIF-2 alpha|hypoxia-inducible factor 2 alpha|Member of PAS protein 2|MOP2HIF2-alpha|endothelial PAS domain protein 1|BHLHE73|PASD2PAS domain-containing protein 2|Hypoxia-inducible factor 2-alpha
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Human HIF-2 alpha, corresponding to amino acids 535-631. [UniProt# Q99814]
- Purification:Protein G purified
- Cat. No.:103368-128
The HIF-2 alpha / EPAS1 Antibody (ep190b) [PE] from Novus Biologicals is a mouse monoclonal antibody to HIF-2 alpha / EPAS1. This antibody reacts with human, mouse, rat, hamster. The HIF-2 alpha / EPAS1 Antibody (ep190b) [PE] has been validated for the following applications: Flow Cytometry.
Type: Primary
Antigen: HIF-2 alpha/EPAS1
Clonality: Monoclonal
Clone: ep190b
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Hamster