Anti-Helios Hamster Monoclonal Antibody (DyLight 350) [clone: 22F6]
103407-616
- Antibody Type:Primary
- Antigen Symbol:Helios
- Clonality:Monoclonal
- Clone:22F6
- Conjugation:DyLight® 350
- Flow Cytometry:Yes
- Host:Hamster
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 μl
- Gene ID:22807
- Antigen Synonyms:HELIOS|zinc finger protein Helios|IKAROS family zinc finger 2 (Helios)|zinc finger DNA binding protein Helios|ZNF1A2|MGC34330|2 (Helios)|subfamily 1A|ZNFN1A2|Ikaros family zinc finger protein 2
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Peptide coresponding to the amino acids 51-107 of Helios.
- Purification:Protein A purified
- Cat. No.:103407-616
The Helios Antibody (22F6) [DyLight 350] from Novus Biologicals is a hamster monoclonal antibody to Helios. This antibody reacts with human, mouse. The Helios Antibody (22F6) [DyLight 350] has been validated for the following applications: Flow Cytometry.
Type: Primary
Antigen: Helios
Clonality: Monoclonal
Clone: 22F6
Conjugation: DyLight 350
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Human, Mouse