Anti-Hck Mouse Monoclonal Antibody [clone: 1D9-1A9]
103327-904
- Antibody Type:Primary
- Antigen Symbol:Hck
- Clonality:Monoclonal
- Clone:1D9-1A9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:3055
- Antigen Synonyms:EC 2.7.10|hemopoietic cell kinaseJTK9|tyrosine-protein kinase HCK|p59-HCK/p60-HCK|EC 2.7.10.2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:HCK (AAH14435, 1 a.a. - 505 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MGCMKSKFLQVGGNTSSKTETSASPHCPVYVPDPTSTIKPGPNSHNSNTPGIREAGSEDIIVVALYDYEAIHHEDLSFQKGDQMVVLEESGEWWKARSLATRKEGYIPSNYVARVDSLETEEWFFKGISRKDAERQLLAPGNMLGSFMIRDSETTKGSYSLSVRDYDPRQGDTVKHYKIRTLDNGGFYISPRSTFSTLQELVDHYKKGNDGLCQKLSVPCMSSKPQKPWEKDAWEIPRESLKLEKKLGAGQFGEVWMATYNKHTKVAVKTMKPGSMSVEAFLAEANVMKTLQHDKLVKLHAVVTKEPIYIITEFMAKGSLLDFLKSDEGSKQPLPKLIDFSAQIAEGMAFIEQRNYIHRDLRAANILVSASLVCKIADFGLARVIEDNEYTAREGAKFPIKWTAPEAINFGSFTIKSDVWSFGILLMEIVTYGRIPYPGVSNPEVIRALERGYRMPRPENCPEELYNIMMRCWKNRPEERPTFEYIQSVLDDFYTATESQYQQQP
- Purification:IgG purified
- Cat. No.:103327-904
- Supplier no.:H00003055-M01
The Hck Antibody (1D9-1A9) from Novus Biologicals is a mouse monoclonal antibody to Hck. This antibody reacts with human. The Hck Antibody (1D9-1A9) has been validated for the following applications: Western Blot, ELISA, Immunoprecipitation.
Type: Primary
Antigen: Hck
Clonality: Monoclonal
Clone: 1D9-1A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human