Anti-HAI-1/HGFA Inhibitor 1 Rabbit Polyclonal Antibody
Catalog # 103279-872
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:HAI-1/HGFA Inhibitor 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:6692
- Antigen Synonyms:Kunitz type 1|HAI|Hepatocyte growth factor activator inhibitor type 1|HAI-1|kunitz-type protease inhibitor 1|serine peptidase inhibitor|serine protease inhibitor|hepatocyte growth factor activator inhibitor 1|MANSC2|HAI1
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:ACFLINCLYEQNFVCKFAPREGFINYLTREVYRSYRQLRTQGFGGSGIPKAWAGIDLKVQPQEPLVLKDVENTDWRLLRGDTDVRVERKDPNQVELWGLKEGTYLFQLTVTSSDHP
- Purification:Immunogen affinity purified
- Cat. No.:103279-872
- Supplier no.:NBP1-89720
Specifications
About this item
The HAI-1 / HGFA Inhibitor 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to HAI-1 / HGFA Inhibitor 1. This antibody reacts with human. The HAI-1 / HGFA Inhibitor 1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: HAI-1/HGFA Inhibitor 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human