Anti-GSTO2 Rabbit Polyclonal Antibody
Catalog # 103346-484
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:GSTO2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:119391
- Antigen Synonyms:glutathione S-transferase omega 2|bA127L20.1 (novel glutathione-S-transferase)|glutathione S-transferase omega-2|bA127L20.1|GSTO-2|EC 2.5.1.18|glutathione-S-transferase-like protein
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:GSTO2 (NP_899062.1, 1 a.a. ~ 243 a.a) full-length human protein.MSGDATRTLGKGSQPPGPVPEGLIRIYSMRFCPYSHRTRLVLKAKDIRHEVVNINLRNKPEWYYTKHPFGHIPVLETSQCQLIYESVIACEYLDDAYPGRKLFPYDPYERARQKMLLELFCKVPHLTKECLVALRCGRECTNLKAALRQEFSNLEEILEYQNTTFFGGTCISMIDYLLWPWFERLDVYGILDCVSHTPALRLWISAMKWDPTVCALLMDKSIFQGFLNLYFQNNPNAFDFGLC
- Purification:Protein A purified
- Cat. No.:103346-484
- Supplier no.:H00119391-D01P
Specifications
About this item
The GSTO2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to GSTO2. This antibody reacts with human. The GSTO2 Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: GSTO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human