Anti-GPR1 Mouse Polyclonal Antibody
Catalog # 103327-440
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:GPR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:2825
- Antigen Synonyms:G protein-coupled receptor 1|probable G-protein coupled receptor 1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:GPR1 (NP_005270.2, 1 a.a. - 355 a.a.) full-length human protein. MEDLEETLFEEFENYSYDLDYYSLESDLEEKVQLGVVHWVSLVLYCLAFVLGIPGNAIVIWFTGFKWKKTVTTLWFLNLAIADFIFLLFLPLYISYVAMNFHWPFGIWLCKANSFTAQLNMFASVFFLTVISLDHYIHLIHPVLSHRHRTLKNSLIVIIFIWLLASLIGGPALYFRDTVEFNNHTLCYNNFQKHDPDLTLIRHHVLTWVKFIIGYLFPLLTMSICYLCLIFKVKKRSILISSRHFWTILVVVVAFVVCWTPYHLFSIWELTIHHNSYSHHVMQAGIPLSTGLAFLNSCLNPILYVLISKKFQARFRSSVAEILKYTLWEVSCSGTVSEQLRNSETKNLCLLETAQ
- Purification:Protein A purified
- Cat. No.:103327-440
- Supplier no.:H00002825-B01P
Specifications
About this item
The GPR1 Antibody from Novus Biologicals is a mouse polyclonal antibody to GPR1. This antibody reacts with human. The GPR1 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: GPR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human