Florisil® 30-60 mesh for column chromatography
103518-958
: Avantor
:
- Pack type:Glass bottle
- Size:500 g
- Cat. No.:103518-958
- Appearance:White Powder (May contain black particles)
- Silicate (as SiO2):83.5 to 84.5 %
- Sodium sulfate (Na2SO4):≤1.0
- Magnesium oxide (MgO):15 to 16 %
:
:100.38 g/mol
:Ambient
:1343-88-0
:MgO₃Si
:MFCD00078249