Order Entry
United States
ContactUsLinkComponent
Dichloromethane 99+% stabilized, extra pure
Irritant
Health
Dichloromethane 99+% stabilized, extra pure
Certificates
undefined
Irritant
Health
Dichloromethane 99+% stabilized, extra pure
Certificates

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.05 % (K.F.)
  • GC:
    ≥99.0 %
  • Stabilizer:
    Approx. 100 ppm Amylene
  • Residue after evaporation:
    ≤0.05 %

Specification Test Results

Related Information

Frequently Bought Together