D-(+)-Glucose monohydrate 99+%, extra pure
Marketsource
About this item
Synonyms: (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
- CAS Number:14431-43-7
- Formula:C₆H₁₂O₆·H₂O
- MDL Number:MFCD00149450
- Molecular Weight:198.17 g/mol
- Storage Temperature:Ambient
0 Options Available Below
Product Details & Documents
Synonyms: (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
- CAS Number:14431-43-7
- Formula:C₆H₁₂O₆·H₂O
- MDL Number:MFCD00149450
- Molecular Weight:198.17 g/mol
- Storage Temperature:Ambient
Specifications for Product Family
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Water:8 to 10 %
- HPLC:≥99.0 % (ELSD)
- Residue after ign. (sulfated):≤0.1 %



