D-(+)-Glucose-6-phosphate sodium salt ≥98%
1 g, Glass bottle
Catalog # 103518-098
Marketsource
D-(+)-Glucose-6-phosphate sodium salt ≥98%
Catalog # 103518-098
Marketsource
CAS Number: 54010-71-8
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
CLINDAMYCIN HYDROCHLORIDE 10MG Catalog #TS44351-0100
Product TS44351-0100 is similar to 103518-098 but manufacturer and specifications may vary.
B-D-GLUCOSE 6-PHOSPHATE 1G Catalog #CH00866-1G
Product CH00866-1G is similar to 103518-098 but manufacturer and specifications may vary.
D(+)-GLUCOSE 6-PHOSPHATE SODIUM SA 1G Catalog #TS44698-0010
Product TS44698-0010 is similar to 103518-098 but manufacturer and specifications may vary.
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Synonyms: ROBISON ESTER MONOSODIUM SALT, Sodium (2R,3R,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexyl hydrogenphosphate, G-6-P Na, BETA-D-GLUCOSE 6-PHOSPHATE MONOSODIUM SALT, D-Glucose 6-phosphate sodium
- CAS Number:54010-71-8
- Formula:C₆H₁₂NaO₉P
- MDL Number:MFCD00006611
- Molecular Weight:282.12 g/mol
- Melting Pt:192…197 °C
- Storage Temperature:Refrigerator
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Water:≤6 %
- Specific optical rotation:+31° to +35° (20 °C, 589 nm) (c=10 in H2O)
- Titration with HClO4:97.5 to 102.5 %
Specifications
- Pack type:Glass bottle
- Size:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



