Agar, Bacteriological Grade
Supplier: Avantor
Certificates
About this item
CAS Number:9002-18-0
:
MDL Number:MFCD00081288
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to cream to light yellow to beige Powder
- Infrared spectrum:Conforms
- Loss on drying:≤15 % (1 g, 105 °C)
- Solubility:(1.5 % in hot water) Clear to slightly opalescent light yellow to yellow solution
- Gel strength:≥700 g/cm² (1.5 %)
- Gel point:32 to 38°C (1.5 %)
- Ash:≤5 %
- pH:6.0 to 7.5 (10 % suspension)
Specification Test Results
Frequently Bought Together