Anti-Genethonin 1 Rabbit Polyclonal Antibody
103279-526
- Antibody Type:Primary
- Antigen Name:Genethonin 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:8987
- Antigen Synonyms:genethonin 1|GENX-3414|starch-binding domain-containing protein 1|FLJ41801|starch binding domain 1|Genethonin-1|GENEX3414
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:EHLQESNGHLISKTKDLGKLQAASWRLQNPSREVCDNSREHVPSGQFPDTEAPATSETSNSRSYSEVSRNESLESPMGEWGFQKGQEISAKAATCFAEKLPSSNLLKNRAKEEMSLSDLNSQDRV
- Purification:Immunogen affinity purified
- Cat. No.:103279-526
- Supplier no.:NBP1-91935
The Genethonin 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Genethonin 1. This antibody reacts with human. The Genethonin 1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Genethonin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human