Anti-Cytochrome C Oxidase subunit 6c Mouse Polyclonal Antibody
Catalog # 103324-218
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Cytochrome C Oxidase subunit 6c
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:1345
- Antigen Synonyms:cytochrome c oxidase subunit VIc preprotein|cytochrome c oxidase subunit VIc|cytochrome c oxidase subunit 6C|Cytochrome c oxidase polypeptide VIc
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:COX6C (NP_004365.1, 1 a.a. - 75 a.a.) full-length human protein. MAPEVLPKPRMRGLLARRLRNHMAVAFVLSLGVAALYKFRVADQRKKAYADFYRNYDVMKDFEEMRKAGIFQSVK
- Purification:Protein G purified
- Cat. No.:103324-218
- Supplier no.:H00001345-B01P
Specifications
About this item
The Cytochrome C Oxidase subunit 6c Antibody from Novus Biologicals is a mouse polyclonal antibody to Cytochrome C Oxidase subunit 6c. This antibody reacts with human. The Cytochrome C Oxidase subunit 6c Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: Cytochrome C Oxidase subunit 6c
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human