Anti-Cystathionase Mouse Monoclonal Antibody [clone: 2E1.2-1C10]
Catalog # 103324-614
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Cystathionase
- Clonality:Monoclonal
- Clone:2E1.2-1C10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:1491
- Antigen Synonyms:homoserine deaminase|EC 4.4.1|MGC9471|homoserine dehydratase|cystathionase (cystathionine gamma-lyase)|EC 4.4.1.1|cysteine desulfhydrase|cystathionine gamma-lyase|gamma-cystathionase
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CTH (AAH15807, 1 a.a. - 405 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MQEKDASSQGFLPHFQHFATQAIHVGQDPEQWTSRAVVPPISLSTTFKQGAPGQHSGFEYSRSGNPTRNCLEKAVAALDGAKYCLAFASGLAATVTITHLLKAGDQIICMDDVYGGTNRYFRQVASEFGLKISFVDCSKIKLLEAAITPETKLVWIETPTNPTQKVIDIEGCAHIVHKHGDIILVVDNTFMSPYFQRPLALGADISMYSATKYMNGRSDVVMGLVSVNCESLHNRLRFLQNSLGAVPSPIDCYLCNRGLKTLHVRMEKHFKNGMAVAQFLESNPWVEKVIYPGLPSHPQHELVKRQCTGCTGMVTFYIKGTLQHAEIFLKNLKLFTLAESLGGFESLAELPAIMTHASVLKNDRDVLGISDTLIRLSVGLEDEEDLLEDLDQALKAAHPPSGSHS
- Purification:IgG purified
- Cat. No.:103324-614
- Supplier no.:H00001491-M02
Specifications
About this item
The Cystathionase Antibody (2E1.2-1C10) from Novus Biologicals is a mouse monoclonal antibody to Cystathionase. This antibody reacts with human, mouse. The Cystathionase Antibody (2E1.2-1C10) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: Cystathionase
Clonality: Monoclonal
Clone: 2E1.2-1C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse