To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Cleavage stimulation factor
- Clonality:Monoclonal
- Clone:1C6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:1477
- Antigen Synonyms:50kD|50kDa|cleavage stimulation factor|subunit 1|CSTF 50 kDa subunit|CstFp50|Cleavage stimulation factor 50 kDa subunit|3' pre-RNA|CF-1 50 kDa subunit|CstF-50|cleavage stimulation factor subunit 1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CSTF1 (NP_001315.1 332 a.a. - 431 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LWEISTGRTLVRYTGAGLSGRQVHRTQAVFNHTEDYVLLPDERTISLCCWDSRTAERRNLLSLGHNNIVRCIVHSPTNPGFMTCSDDFRARFWYRRSTTD
- Purification:IgG purified
- Cat. No.:103324-562
- Supplier no.:H00001477-M01
Specifications
About this item
The cleavage stimulation factor Antibody (1C6) from Novus Biologicals is a mouse monoclonal antibody to cleavage stimulation factor. This antibody reacts with human. The cleavage stimulation factor Antibody (1C6) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: Cleavage stimulation factor
Clonality: Monoclonal
Clone: 1C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human