Anti-Claudin-19 Mouse Monoclonal Antibody [clone: 2F2]
Catalog # 103347-238
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Claudin-19
- Clonality:Monoclonal
- Clone:2F2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:149461
- Antigen Synonyms:claudin-19|claudin 19
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CLDN19 (AAH30524, 1 a.a. ~ 212 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.MANSGLQLLGYFLALGGWVGIIASTALPQWKQSSYAGDAIITAVGLYEGLWMSCASQSTGQVQCKLYDSLLALDGHIQSARALMVVAVLLGFVAMVLSVVGMKCTRVGDSNPIAKGRVAIAGGALFILAGLCTLTAVSWYATLVTQEFFNPSTPVNARYEFGPALFVGWASAGLAVLGGSFLCCTCPEPERPNSSPQPYRPGPSAAAREYV
- Purification:IgG purified
- Cat. No.:103347-238
- Supplier no.:H00149461-M02
Specifications
About this item
The Claudin-19 Antibody (2F2) from Novus Biologicals is a mouse monoclonal antibody to Claudin-19. This antibody reacts with human, mouse. The Claudin-19 Antibody (2F2) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: Claudin-19
Clonality: Monoclonal
Clone: 2F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse