Anti-GALNT6 Rabbit Polyclonal Antibody
103272-538
- Antibody Type:Primary
- Antigen Symbol:GALNT6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:11226
- Antigen Synonyms:GalNAc transferase 6|GALNAC-T6|polypeptide N-acetylgalactosaminyltransferase 6|EC 2.4.1.41|Polypeptide GalNAc transferase 6|UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 6|pp-GaNTase 6|UDP-N-acetyl-alpha-D-galactosamine: polypeptideN-acetylgalactosaminyltransferase 6|UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 6 (GalNAc-T6)|GalNAcT6|Protein-UDP acetylgalactosaminyltransferase 6
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:LIMYSCHGLGGNQYFEYTTQRDLRHNIAKQLCLHVSKGALGLGSCHFTGKNSQVPKDEEWELAQDQLIRNSGSGTCLTSQDKKPAMAPCNPSDPHQLWLFV
- Purification:Immunogen affinity purified
- Cat. No.:103272-538
- Supplier no.:NBP1-86100
The GALNT6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to GALNT6. This antibody reacts with human. The GALNT6 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: GALNT6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human