Anti-GADD45 alpha Mouse Monoclonal Antibody [clone: 3D12]
103324-982
- Antibody Type:Primary
- Antigen Symbol:GADD45 alpha
- Clonality:Monoclonal
- Clone:3D12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:1647
- Antigen Synonyms:growth arrest and DNA-damage-inducible|alpha|GADD45DNA-damage-inducible transcript 1|DDIT-1|DNA damage-inducible transcript 1 protein|growth arrest and DNA-damage-inducible 45 alpha|DDIT1growth arrest and DNA damage-inducible protein GADD45 alpha|DNA damage-inducible transcript-1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:GADD45A (AAH11757 76 a.a. - 165 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. TLIQAFCCENDINILRVSNPGRLAELLLLETDAGPAASEGAEQPPDLHCVLVTNPHSSQWKDPALSQLICFCRESRYMDQWVPVINLPER
- Purification:IgG purified
- Cat. No.:103324-982
- Supplier no.:H00001647-M01
The GADD45 alpha Antibody (3D12) from Novus Biologicals is a mouse monoclonal antibody to GADD45 alpha. This antibody reacts with human. The GADD45 alpha Antibody (3D12) has been validated for the following applications: Western Blot, ELISA, Proximity Ligation Assay.
Type: Primary
Antigen: GADD45 alpha
Clonality: Monoclonal
Clone: 3D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human