Anti-GABPA Mouse Monoclonal Antibody [clone: 4B8-G6]
103326-892
- Antibody Type:Primary
- Antigen Symbol:GABPA
- Clonality:Monoclonal
- Clone:4B8-G6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:2551
- Antigen Synonyms:E4TF1-60|human nuclear respiratory factor-2 subunit alpha|Nuclear respiratory factor 2 subunit alpha|GA-binding protein alpha chain|alpha subunit (60kD)|nuclear respiratory factor 2 alpha subunit|NRF2|NRF2A|E4TF1ATranscription factor E4TF1-60|alpha subunit 60kDa|GA binding protein transcription factor|GA-binding protein transcription factor|GABP subunit alpha|10|NFT2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:GABPA (AAH35031, 1 a.a. - 454 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MTKREAEELIEIEIDGTEKAECTEESIVEQTYAPAECVSQAIDINEPIGNLKKLLEPRLQCSLDAHEICLQDIQLDPERSLFDQGVKTDGTVQLSVQVISYQGIEPKLNILEIVKPADTVEVVIDPDAHHAESEAHLVEEAQVITLDGTKHITTISDETSEQVTRWAAALEGYRKEQERLGIPYDPIQWSTDQVLHWVVWVMKEFSMTDIDLTTLNISGRELCSLNQEDFFQRVPRGEILWSHLELLRKYVLASQEQQMNEIVTIDQPVQIIPASVQSATPTTIKVINSSAKAAKVQRAPRISGEDRSSPGNRTGNNGQIQLWQFLLELLTDKDARDCISWVGDEGEFKLNQPELVAQKWGQRKNKPTMNYEKLSRALRYYYDGDMICKVQGKRFVYKFVCDLKTLIGYSAAELNRLVTECEQKKLAKMQLHGIAQPVTAVALSTASLQTEKDN
- Purification:IgG purified
- Cat. No.:103326-892
- Supplier no.:H00002551-M01
The GABPA Antibody (4B8-G6) from Novus Biologicals is a mouse monoclonal antibody to GABPA. This antibody reacts with human. The GABPA Antibody (4B8-G6) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: GABPA
Clonality: Monoclonal
Clone: 4B8-G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human