To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:FoxP3
- Clonality:Monoclonal
- Clone:3G3
- Conjugation:DyLight® 650
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:50943
- Antigen Synonyms:immune dysregulation|enteropathy|Enteropathy|Polyendocrinopathy|AIID|FOXP3delta7|PIDX|JM2|forkhead box P3|MGC141961|X-linked|XPIDpolyendocrinopathy|X-Linked|Forkhead Box Protein P3|PIDXMGC141963|DIETER|XPID|AIIDMGC141961|Immunodeficiency|Scurfin|polyendocrinopathy|MGC141963|IPEX
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Full-length His-tagged murine FOXP3 was used as the immunogen. The precise epitope is not known, but it has been mapped to the N-terminal portion of the protein (NP_001107849).
- Purification:Protein G purified
- Cat. No.:103360-458
Specifications
About this item
The FoxP3 Antibody (3G3) [DyLight 650] from Novus Biologicals is a mouse monoclonal antibody to FoxP3. This antibody reacts with human, mouse. The FoxP3 Antibody (3G3) [DyLight 650] has been validated for the following applications: Western Blot, Flow Cytometry.
Type: Primary
Antigen: FoxP3
Clonality: Monoclonal
Clone: 3G3
Conjugation: DyLight 650
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse