Anti-FOXD4 Mouse Polyclonal Antibody
Catalog # 103326-524
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:FOXD4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:2298
- Antigen Synonyms:FREAC-5|homolog-like 9|Forkhead-related transcription factor 5|Drosophila|FREAC5FOXD4a|forkhead|forkhead-like 9|forkhead-related activator 5|FKHL9MGC105106|forkhead box D4|FOXD4A|forkhead box protein D4|Forkhead-related protein FKHL9|Myeloid factor-alpha
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:FOXD4 (NP_997188.2, 1 a.a. - 439 a.a.) full-length human protein. MNLPRAERLRSTPQRSLRDSDGEDGKIDVLGEEEDEDEEEAASQQFLEQSLQPGLQVARWGGVALPREHIEGGGGPSDPSEFGTEFRAPPRSAAASEDARQPAKPPSSYIALITMAILQSPHKRLTLSGICAFISDRFPYYRRKFPAWQNSIRHNLSLNDCFVKIPREPGRPGKGNYWSLDPASQDMFDNGSFLRRRKRFQRHQPTPGAHLPHPFPLPAAHAALHNPRPGPLLGAPAPPQPVPGAYPNTGPGRRPYALLHPHPPRYLLLSAPAYAGAPKKAEGADLATPAPFPCCSPHLVLSLGRRARVWRRHREADASLSALRVSCKGSGERVQGLRRVCPRPRGATAPCSSDRQACRTILQQQQRHQEEDCANGCAPTKGAVLGGHLSAASALLRYQAVAEGSGLTSLAAPLGGEGTSPVFLVSPTPSSLAESAGPS
- Purification:Protein A purified
- Cat. No.:103326-524
- Supplier no.:H00002298-B01P
Specifications
About this item
The FOXD4 Antibody from Novus Biologicals is a mouse polyclonal antibody to FOXD4. This antibody reacts with human. The FOXD4 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: FOXD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human