Anti-FGL2 Mouse Monoclonal Antibody [clone: 6D9]
103339-714
- Antibody Type:Primary
- Antigen Name:Fibroleukin
- Antigen Symbol:FGL2
- Clonality:Monoclonal
- Clone:6D9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:10875
- Antigen Synonyms:Fibrinogen-like protein 2|fibroleukin|pT49T49|fibrinogen-like 2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:FGL2 (NP_006673, 24 a.a. ~ 123 a.a) partial recombinant protein with GST tag.NNETEEIKDERAKDVCPVRLESRGKCEEAGECPYQVSLPPLTIQLPKQFSRIEEVFKEVQNLKEIVNSLKKSCQDCKLQADDNGDPGRNGLLLPSTGAPG
- Purification:Protein A purified
- Cat. No.:103339-714
- Supplier no.:H00010875-M01
The FGL2 / Fibroleukin Antibody (6D9) from Novus Biologicals is a mouse monoclonal antibody to FGL2 / Fibroleukin. This antibody reacts with human, mouse, rat. The FGL2 / Fibroleukin Antibody (6D9) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: FGL2
Clonality: Monoclonal
Clone: 6D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat